Learn More
MilliporeSigma™ anti-NG2 Chondroitin Sulfate Proteoglycan , ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB5384/DEL

Description
NG2 is a high molecular weight, integral membrane chondroitin sulfate proteoglycan. It is found on the surfaces of an unusual class of glial cells within the developing and mature central nervous system that have the properties of oligodendrocyte precursor cells (i.e., O-2A progenitor cells). NG2 is also found on the surfaces of chondroblasts, proliferating capillary endothelial cells, some human melanoma cell lines, and on leukemic blasts in childhood acute lymphoblastic leukemia. The NG2 proteoglycan is likely to play a role in regulation of cell motility, axon outgrowth and the cellular responses to platelet-derived growth factor.
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blotting

Specifications
NG2 Chondroitin Sulfate Proteoglycan | |
Unconjugated | |
Cell line expressing a truncated form of NG2. | |
RUO | |
Primary | |
Semi-purified |
Monoclonal | |
Mouse | |
100 μg | |
Neuroscience | |
Human, Mouse, Rat | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.