Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NG2 Chondroitin Sulfate Proteoglycan Rabbit anti-Human, Mouse, Rat, Monkey, Polyclonal, MilliporeSigma™

Catalog No. AB5320MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5320MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5320MI Supplier MilliporeSigma Supplier No. AB5320
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect NG2 Chondroitin Sulfate Proteoglycan using this Anti-NG2 Chondroitin Sulfate Proteoglycan Antibody validated for use in ELISA, immunocytochemistry, immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NG2 Chondroitin Sulfate Proteoglycan
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in PBS buffer containing 0.02% azide.
Gene Symbols CSPG4; MSK16; MCSP; NG2; MCSPG; MEL-CSPG; HMW-MAA
Host Species Rabbit
Immunogen Immunoaffinity purified NG2 Chondroitin Sulfate Proteoglycan from rat.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001897.4
Target Species Human, Monkey, Mouse, Rat
Content And Storage -20°C in undiluted aliquotes for up to 6 months, Avoid Freeze/Thaw Cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.