Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NG2 Mouse anti-Mouse, Rat, Clone: 132.38, MilliporeSigma™

Catalog No. 05710MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-710-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-710-MI Supplier MilliporeSigma Supplier No. 05710
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-NG2 Antibody, clone 132.38 detects level of NG2 and has been published and validated for use in immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NG2
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 132.38
Conjugate Unconjugated
Formulation Protein A Purified immunoglobulin in 0.1M Tris-glycine, pH 7.4, 0.15M NaCl with 0.05% sodium azide as a preservative.
Gene Accession No. NM_001897
Gene Symbols CSPG4, MCSP, HMW-MAA, MCSPG, MSK16, NG2, MEL-CSPG
Host Species Mouse
Immunogen HEK293 cells expressing a truncated integral membrane form of NG2 consisting of amino acids 1592-2222
Purification Method Protein G purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.