Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-NG2/MCSP, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP121361
Description
NG2/MCSP Polyclonal antibody specifically detects NG2/MCSP in Human, Porcine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
NG2/MCSP | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 5-10 ug/mg lysate | |
Q6UVK1 | |
chondroitin sulfate proteoglycan 4, chondroitin sulfate proteoglycan 4 (melanoma-associated), EC 3.6.3, HMW-MAA, MCSPChondroitin sulfate proteoglycan NG2, MCSPGMelanoma chondroitin sulfate proteoglycan, MEL-CSPG, MSK16Melanoma-associated chondroitin sulfate proteoglycan, NG2EC 2.7.8 | |
Rabbit | |
IgG | |
0.1 ml | |
Store at 4C. Do not freeze. | |
Polyclonal | |
Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Gorilla, Chimpanzee, and Northern white-cheeked gibbon. |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
Affinity Purified | |
CSPG4 | |
The immunogen recognized by this antibody maps to a region between residue 575 and 625 of human melanoma-associated chondroitin sulfate proteoglycan (chondroitin sulfate proteoglycan 4 ) using the numbering given in entry NP_001888.2 (GeneID 1464). | |
Immunogen affinity purified | |
RUO | |
Primary | |
1464 | |
Human, Porcine |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only