Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NG2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA517199
Description
It is not recommended to aliquot this antibody.
A human melanoma-associated chondroitin sulfate proteoglycan plays a role in stabilizing cell-substratum interactions during early events of melanoma cell spreading on endothelial basement membranes. CSPG4 represents an integral membrane chondroitin sulfate proteoglycan expressed by human maligt melanoma cells.
Specifications
NG2 | |
Polyclonal | |
CSPG4 | |
chondroitin sulfate proteoglycan 4 (melanoma-associated); chondroitin sulfate proteoglycan NG2; melanoma chondroitin sulfate proteoglycan; melanoma-associated chondroitin sulfate proteoglycan | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
121021, 1464, 81651 | |
-20°C | |
Liquid |
Western Blot | |
Unconjugated | |
Q6UVK1, Q8VHY0, Q00657 | |
CSPG4 | |
Synthetic peptide surrounding His2260 of human NG2 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction