Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NHE-1 (Na+/H+ Exchangers) Mouse, Unlabeled, Clone: 54, BD

Catalog No. BDB611774 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611774 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611774 Supplier BD Biosciences Supplier No. 611774

Mouse Monoclonal Antibody

The extrusion of H+ in exchange for extracellular Na+ is important for many cellular processes, such as pH homeostasis, volume regulation, and transepithelial ion and water transport. Na+/H+ Exchangers (NHE) are integral membrane proteins that mediate electroneutral exchange of one Na+ ion for one H+ ion. Six NHE forms, NHE-1 thru -6, have been identified. NHE-1 and NHE-6 are widely expressed, while the other NHE forms have restricted expression. The common structure of all NHE forms includes 10-12 N-terminal membrane (M) spanning regions, a conserved M6 and M7 region that may participate in ion transport, and a large C-terminal cytoplasmic region that may be involved in the regulation of ion exchange activity. NHE-1, for example, contains 12 M regions plus domains for volume sensitivity, calmodulin-binding, CHP-binding, and PKC phosphorylation in the cytoplasmic region. Regulation of NHE-1 ion exchange activity may occur through phosphoinositide binding, as well as PKC- and PKA-dependent signaling pathways. Mutation of NHE-1 in mice causes neuronal death in the cerebellum and brainstem, leading to ataxia and seizures. Thus, NHE-1 is a ubiquitous NHE that is essential for normal brain function.

Although this antibody was developed against the NHE-1 antigen, investigators should note that crossreactivity to other NHE isoforms or variants may be possible.

Although this antibody was developed against the NHE-1 antigen, investigators should note that crossreactivity to other NHE isoforms or variants may be possible.

TRUSTED_SUSTAINABILITY

Specifications

Antigen NHE-1 (Na+/H+ Exchangers)
Applications Western Blot
Classification Monoclonal
Clone 54
Concentration 250μg/mL
Conjugate Unconjugated
Description Na+/H+ Exchangers
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat NHE-1 aa. 682-801
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.