Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NHERF-2 Mouse anti-Human, Rat, Clone: 32B6, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABN385
Description
Anti-NHERF-2 Antibody, clone 32B6 is a highly specific mouse monoclonal antibody, that targets NHERF2 and has been tested in western blotting, immunocytochemistry and immunoprecipitation.
Specifications
NHERF-2 | |
Monoclonal | |
Unconjugated | |
NP_001123484 | |
Mouse | |
Protein G purified | |
Primary | |
2-8°C for one year | |
IgG1 |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
32B6 | |
Purified mouse monoclonal IgG1 in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
SLC9A3R2, NHERF2 | |
recombinant protein corresponding to human NHERF-2. | |
100 μg | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction