Learn More
NHP CD4 Mouse anti-Human, Brilliant Violet 510, Clone: L200, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 563094
Description
The L200 monoclonal antibody specifically binds to the human form of the 56 kDa transmembrane glycoprotein, CD4, present on the T-helper/inducer subset of normal human donor peripheral blood lymphocytes. The L200 antibody also crossreacts with a subset of CD3-positive peripheral blood lymphocytes, but not monocytes, of both Rhesus and Cynomolgus Macaque monkeys. Crossreactivity on both lymphocytes and monocytes (weak) from Baboons is also observed. The distribution on lymphocytes is similar for both human and monkey cells, with the majority of CD4-positive lymphocytes being CD8-negative and lacking reactivity with antibodies to B- or NK-cell markers.
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 405-nm and Em Max at 510-nm, BD Horizon™ BV510 can be excited by the violet laser and detected in the BD Horizon™ V500 (525/50-nm) filter set. BD Horizon™ BV510 conjugates are useful for the detection of dim markers off the violet laser.

Specifications
NHP CD4 | |
Monoclonal | |
Brilliant Violet 510 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human HPB-ALL Cell Line | |
50 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
L200 | |
L3T4 ; T-cell surface antigen T4/Leu-3; W3/25 ; CD4 antigen (p55) | |
Mouse | |
Affinity Purified | |
RUO | |
Baboon, Cynomolgus, Rhesus Monkey | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.