Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Nibrin/Nbs1 Clone: EE15,

Catalog No. 05616CMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
05-616-CMI 200 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-616-CMI Supplier MilliporeSigma™ Supplier No. 05616C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Nibrin/Nbs1 Clone: EE15 in Human samples, and it is validated for Immunocytochemistry, Immunoprecipitation, Western Blotting

Nibrin (UniProt O60934; also known as Cell cycle regulatory protein p95, Nijmegen breakage syndrome protein 1, p95 protein of the MRE11/RAD50 complex) is encoded by the NBN (also known as AT-V1, AT-V2, ATV, NBS, NBS1, P95) gene (Gene ID 4683) in human. Nibrin is a component of the MRN (MRE11/RAD50/NBN) complex involved in early steps of DNA double strand breaks (DSBs). The phosphorylation of H2AX pSer139 (γ-H2AX) found at the DNA-damaged sites, for example, is mediated by a direct interaction between Nibrin and the phosphorylated mediator of the DNA damage checkpoint 1 (MDC1). ATM, CtIP/RBBP8, Tip60, BRCA1, and SMC1 are also known interaction partners of Nibrin. Nibrin contains a fork-head associated (FHA) domain and two BRCA1 C-terminal (BRCT) tandem domains in its N-terminal region, followed by two MRE11-binding motifs and an ATM-binding motif in its C-terminal region. The Ser278 and Ser343 residues are known to be targeted for phosphorylation by ATM in response to ionizing-radiation (IR). NBN gene mutations are responsible for Nijmegen breakage syndrome (NBS), a disorder characterized by chromosomal instability, radiation sensitivity, microcephaly, growth retardation, immunodeficiency and predisposition to cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nibrin/Nbs1
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone EE15
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. O60934
Gene Symbols NBN, NBS, NBS1, P95
Host Species Mouse
Immunogen GST fusion protein corresponding to residues 420-774 of human Nibrin/Nbs1.
Purification Method Protein G purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Epigenetics, Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_002476
Test Specificity Recognizes Nibrin/Nbs1, Mr 95kDa. A non-specific protein was also detected at Mr 33kDa in some lysates, but not Jurkat cell lysate.
Target Species Human
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.