Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicastrin Mouse, Unlabeled, Clone: 35, BD

Catalog No. BDB612290 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612290 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612290 Supplier BD Biosciences Supplier No. 612290

Mouse Monoclonal Antibody

Amyloid precursor protein (APP) gene encodes multiple APPs ranging from 695 to 770 amino acids. The unprocessed form of APP is a putative cell surface receptor that possesses neurite-promoting activity, and is involved in synaptic vesicle recycling. Processing of APP by sequential enzymatic activity of β- and γ-secretase, and the presenilin proteins PS1 and PS2, produces APP fragments that may have unique functions. β-secretase activity cleaves the extracellular portion of APP leading to asecreted APP form, while γ-secretase activity produces β-Amyloid peptide (39-43 amino acids). The Aβ peptide produces abnormal plaques in the cerebral cortex and blood vesselwalls during Alzheimer's disease. Nicastrin is a PS1 associated transmembrane protein,that contains N-terminal glycosylation and myristoylation sites. Nicastrin can bind full length APP and the fragments produced by γ-secretase. In C. elegans, suppression of nicastrin expression produces phenotypes that mimic those produced when notch signaling proteins are suppressed. In Drosophila, deficiencies in nicastrin prevent cleavage of the intracellular portion of Notch. Thus, nicastrin may be a functional component of presenilins and γ-secretase complexes, which process Notch and APP transmembrane receptors.

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Nicastrin
Applications Western Blot
Classification Monoclonal
Clone 35
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Nicastrin aa. 168-289
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.