Learn More
Nicotinic Acetylcholine Receptor α4 Rabbit anti-Mouse, Rat, Human, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Nicotinic Acetylcholine Receptor α4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Rabbit purified polyclonal serum in buffer containing 0.1M Tris-Glycine (pH7.4) 150mM NaCl with 0.05% sodium azide. |
| Gene Symbols | ACHRA; ACHRD; CHNRA; CHRNA; CMS2A; FCCMS; SCCMS |
| Host Species | Rabbit |
| Immunogen | This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from residues of Nicotinic Acetylcholine Receptor α4. |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.