Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nidogen 1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016862 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-862 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-862 Supplier Abnova Corporation Supplier No. H00004811B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human NID1 protein.

This gene encodes a member of the nidogen family of basement membrane glycoproteins. The protein interacts with several other components of basement membranes, and may play a role in cell interactions with the extracellular matrix. [provided by RefSeq

Sequence: MLASSSRIRAAWTRALLLPLLLAGPVGCLSRQELFPFGPGQGDLELEDGDDFVSPALELRGALRFYDRSDIDAVYVTTNGIIATSEPPAKESHPGLFPPTFGAVAPFLADLDTTDGLGKVYYREDLSPSITQRAAECVHRGFPEISFQPSSAVVVTWESVAPYQGPSRDPDQKGKRNTFQAVLASSDSSSYAIFLYPEDGLQFHTTFSKKENNQVPAVVAFSQGSVGFLWKSNGAYNIFANDRESIENLAKSSNSGQQGVWVFEIGSPATTNGVVPADVILGTEDGAEYDDEDEDYDLATTRLGLEDVGTTPFSYKALRRGGADTYSVPSVLSPRRAATERPLGPPTERTRSFQLAVETFHQQHPQVIDVDEVEETGVVFSYNTDSRQTCANNRHQCSVHAECRDYATGFCCSCVAGYTGNGRQCVAEGSPQRVNGKVKGRIFVGSSQVPIVFENTDLHSYVVMNHGRSYTAISTIPETVGYSLLPLAPVGGIIGWMFAVEQDGFKNGFSITGGEFTRQAEVTFVGHPGNLVIKQRFSGIDEHGHLTIDTELEGRVPQIPFGSSVHIEPYTELYHYSTSVITSSSTREYTVTEPERDGASPSRIYTYQWRQTITFQECVHDDSRPALPSTQQLSVDSVFVLYNQEEKILRYALSNSIGPVREGSPDALQNPCYIGTHGCDTNAACRPGPRTQFTCECSIGFRGDGRTCYEVEKTRCQHEREHILGAAGATDPQRPIPPGLFVPECDAHGHYAPTQCHGSTGYCWCVDRDGREVEGTRTRPGMTPPCLSTVAPPIHQGPAVPTAVIPLPPGTHLLFAQTGKIERLPLEGNTMRKTEAKAFLHVPAKVIIGLAFDCVDKMVYWTDITEPSIGRASLHGGEPTTIIRQDLGSPEGIAVDHLGRNIFWTDSNLDRIEVAKLDGTQRRVLFETDLVNPRGIVTDSVRGNLYWTDWNRDNPKIETSYMDGTNRRILVQDDLGLPNGLTFDAFSSQLCWVDAGTNRAECLNPSQPSRRKALEGLQYPFAVTSYGKNLYFTDWKMNSVVALDLAISKETDAFQPHKQTRLYGITTALSQCPQGHNYCSVNNGGCTHLCLATPGSRTCRCPDNTLGVDCIEQK

Specifications

Antigen nidogen 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human NID1 protein.
Formulation PBS with no preservative; pH 7.4
Gene NID1
Gene Accession No. BC045606.1
Gene Alias NID
Gene Symbols NID1
Host Species Mouse
Immunogen NID1 (AAH45606.1, 1 a.a. ∼ 1114 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4811
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.