Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nidogen Rat anti-Human, Mouse, Clone: JF6, MilliporeSigma™

Catalog No. MAB1886MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1886MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1886MI Supplier MilliporeSigma Supplier No. MAB1886

Rat Monoclonal Antibody

Detect Nidogen using this Anti-Nidogen Antibody, G1 domain, clone JF6 validated for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nidogen
Applications ELISA, Western Blot
Classification Monoclonal
Clone JF6
Conjugate Unconjugated
Formulation Liquid at 1mg/mL in 0.02 MPB + 0.25MNaCl, containing 0.1% sodium azide as preservative. pH 7.6
Gene Accession No. NM_002508.2
Gene Symbols NID1, enactin, NID, Entactin, entactin
Host Species Rat
Immunogen Recombinant fragment corresponding to the G1 region of mouse nidogen.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.