Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ninjurin Mouse, Unlabeled, Clone: 50, BD

Catalog No. BDB610777 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610777 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610777 Supplier BD Biosciences Supplier No. 610777
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ninjurin is a protein whose expression is dramatically increased after sciatic nerve transection and crush injuries. The Ninjurin gene encodes for a protein of 152 amino acids with two putative transmembrane domains. Ninjurin was iodinated in vivo and promoted the aggregation of Jurkat cells expressing Ninjurin, indicating a portion of Ninjurin is exposed to the cell surface. The adhesive properties of Ninjurin were energy and temperature dependent, required Ca2+; and Mg2+, and the integrity of the cytoskeleton. Also, the adhesion domain of Ninjurin is located at the extracellular NH2-terminal domain. Although its predicted mass is 16kDa, Ninjurin migrates as a 18-22kDa protein in SDS-PAGE, depending on the cell line or tissue. mRNA analysis revealed that Ninjurin is widely expressed with the highest levels in liver, thymus, heart, and the lowest level in brain.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ninjurin
Applications Western Blot
Classification Monoclonal
Clone 50
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Ninjurin aa. 1-152
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.