Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrated Tau (Tyr29) Mouse anti-Human, Clone: Tau-nY29, MilliporeSigma™

Catalog No. MAB2244MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2244MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2244MI Supplier MilliporeSigma Supplier No. MAB2244

Mouse Monoclonal Antibody

Detect Nitrated Tau (Tyr29) using this Anti-Nitrated Tau (Tyr29) Antibody, clone Tau-nY29 validated for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nitrated Tau (Tyr29)
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone Tau-nY29
Conjugate Unconjugated
Formulation Purified in 0.1M Tris-Glycine (pH7.4) 150mM NaCl with 0.05% NaN3.
Gene Accession No. P10636
Gene Symbols DDPAC , FLJ31424, FTDP-17 , MAPTL, MSTD , MTBT1, MTBT2 , PHF-tau , PPND , TAU, tau
Host Species Mouse
Immunogen KLH conjugated synthetic linear peptide corresponding to the amino acids surrounding and including nitrated Tyr29 of human Tau.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.