Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Nitric Oxide Synthase (1131-1144), Inducible, Polyclonal

Catalog No. 482728100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
48-272-8100UL 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-272-8100UL Supplier MilliporeSigma™ Supplier No. 482728100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Nitric Oxide Synthase (1131-1144), Inducible Clone: in Human, Mouse, Rat samples, and it is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immuno Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Nitric Oxide Synthase (1131-1144), Inducible
Applications Immunoblot, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation Undiluted serum.
Host Species Rabbit
Immunogen a synthetic peptide (CKKGSALEERKATRL) corresponding to amino acids 1131-1144 of mouse macrophage inducible nitric oxide synthase (iNOS), conjugated to KLH
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽130kDa iNOS protein.
Target Species Human, Mouse, Rat
Content And Storage −20°C
Form Undiluted
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.