Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Oxide Synthase I Rabbit anti-Bovine, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB1632MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1632MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1632MI Supplier MilliporeSigma Supplier No. AB1632
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Nitric Oxide Synthase I Antibody is an antibody against Nitric Oxide Synthase I for use in ELISA, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nitric Oxide Synthase I
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Peptide affinity-purified immunoglobulin. Supplied at a 1mg/mL solution in PBS, pH 7.4, containing 0.1% BSA.
Gene Symbols NOS1; nNOS; NC-NOS; N-NOS; IHPS1; bNOS; NOS; EC 1.14.13.39
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 724-739 of human brain NOS, coupled to KLH.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_000620.1
Target Species Bovine, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.