Learn More
Nitric Oxide Synthase I Sheep anti-Guinea Pig, Rat, Rabbit, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Nitric Oxide Synthase I |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Sheep serum. Lyophilized, no preservatives. Reconstitute with 100μL of sterile distilled water. Centrifuge to remove any residue. |
| Gene Symbols | NOS1; nNOS; NC-NOS; N-NOS; IHPS1; bNOS; NOS; EC 1.14.13.39 |
| Host Species | Sheep |
| Immunogen | A 21 amino acid peptide (1409-1429) of the neuronal form of NOS from rat cerebellum, coupled to KLH. The sequence is H-RSESIAFIEESKKDADEVFSS-NH2. |
| Quantity | 100 μL |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.