Learn More
Nitric Oxide Synthase II Rabbit anti-Mouse, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Nitric Oxide Synthase II |
| Applications | Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in 0.02M Phosphate buffer, 0.25M NaCl, pH 7.6 with 0.1% Sodium azide as a preservative. |
| Gene Symbols | NOS2A; iNOS; HEP-NOS; INOS; NOS; NOS2; EC 1.14.13.39 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide corresponding to amino acids 1131-1144 of mouse macrophage NOS, coupled to KLH. |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.