Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Oxide Synthase II Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB5384MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5384MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5384MI Supplier MilliporeSigma Supplier No. AB5384
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Nitric Oxide Synthase II using this Anti-Nitric Oxide Synthase II Antibody validated for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nitric Oxide Synthase II
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit serum. Liquid containing 0.05% sodium azide.
Gene Symbols NOS2A; iNOS; HEP-NOS; INOS; NOS; NOS2; EC 1.14.13.39
Host Species Rabbit
Immunogen C-terminal 19 amino acid peptide from human chondrocyte nitric oxide synthase.
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_153292.1; NM_000625.3
Target Species Human
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.