Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitro-α/Β-Synuclein Mouse anti-Human, Clone: nSyn12, MilliporeSigma™

Catalog No. 36011MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-011-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-011-MI Supplier MilliporeSigma Supplier No. 36011
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-nitro-α/β-Synuclein Antibody, clone nSyn12 is an antibody against nitro-α/β-Synuclein for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen nitro-α/β-Synuclein
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone nSyn12
Conjugate Unconjugated
Formulation ascites with 0.05% sodium azide
Gene Accession No. NM_000345.2; NM_007308.1
Gene Symbols SNCA, MGC110988, PD1, alpha-synuclein, NACP, PARK1, PARK4, Alpha-synuclein
Host Species Mouse
Immunogen full-length recombinant human α-Synuclein nitrated in vitro
Purification Method Ascites
Quantity 200 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C for 24 months.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.