Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrotyrosine Rabbit anti-All, Polyclonal, MilliporeSigma™

Catalog No. AB5411MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5411MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5411MI Supplier MilliporeSigma Supplier No. AB5411
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Nitrotyrosine using this Anti-Nitrotyrosine Antibody validated for use in immunohistochemistry (paraffin) and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nitrotyrosine
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1% sodium azide.
Host Species Rabbit
Immunogen Nitrated KLH.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species All species
Content And Storage 2-8°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.