Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nitrotyrosine Rabbit anti-All, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 06284
Description
This Anti-Nitrotyrosine Antibody is published in more than 80 citations and validated for use in Immunocytochemistry (IC), Immunohistochemistry (immunohistochemistry), Immunoprecipitation (IP), Western Blotting (WB) for the detection of Nitrotyrosine.
Specifications
Nitrotyrosine | |
Polyclonal | |
Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide as a preservative. | |
Nitrated KLH | |
100 μg | |
All species | |
Purified |
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Immunoaffinity purified | |
Primary | |
−20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction