Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrotyrosine Mouse anti-Vertebrates, AlexaFluor™488, Clone: 1A6, MilliporeSigma™

Catalog No. 16226MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-226-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-226-MI Supplier MilliporeSigma Supplier No. 16226

Mouse Monoclonal Antibody

This Anti-Nitrotyrosine Antibody, clone 1A6, Alexa Fluor 488 conjugate is validated for use in flow cytometry, western blotting for the detection of Nitrotyrosine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nitrotyrosine
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 1A6
Conjugate Alexa Fluor 488
Formulation Protein G Purified mouse monoclonal in a solution of 1% BSA, PBS, 0.05% Tween, and 0.05% sodium azide as a preservative.
Host Species Mouse
Immunogen Nitrated KLH
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Vertebrates
Content And Storage 4°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.