Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrotyrosine Mouse anti-Vertebrates, Clone: 1A6, MilliporeSigma™

Catalog No. 05233MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-233-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-233-MI Supplier MilliporeSigma Supplier No. 05233
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Nitrotyrosine Antibody, clone 1A6 detects level of Nitrotyrosine and has been published and validated for use in immunocytochemistry, immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nitrotyrosine
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1A6
Conjugate Unconjugated
Formulation Protein G Purified immunoglobulin in Protein G Purified immunoglobulin in 0.014M phosphate buffer, pH 7.6, 0.175M NaCl, 0.07% sodium azide, and 30% glycerol.
Host Species Mouse
Immunogen Nitrated KLH
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Vertebrates
Content And Storage −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.