Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nitrotyrosine Mouse anti-All, Clone: 2A8.2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB5404
Description
Detect Nitrotyrosine using this Anti-Nitrotyrosine Antibody, clone 2A8.2 validated for use in immunohistochemistry (paraffin) and western blotting.
Specifications
Nitrotyrosine | |
Monoclonal | |
Unconjugated | |
Mouse | |
Protein A purfied | |
Primary | |
2-8°C for one year | |
IgG2b |
Immunohistochemistry, Western Blot | |
2A8.2 | |
Purified mouse monoclonal IgG2b liquid in buffer containing 0.1% sodium azide. | |
Nitrated KLH. | |
100 μg | |
All species | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction