Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Nitrotyrosine Clone: CC22.8C7.3,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ 48792350UG
Description
Specifically detects Nitrotyrosine Clone: CC22.8C7.3 in samples, and it is validated for Immuno Blotting
Nitric oxide (NO) is a product of the enzymatic conversion of arginine to citrulline by Nitric oxide synthase. The reaction of NO with superoxide which is often present under conditions of oxidative stress, forms peroxynitrite, a strong oxidant which can nitrosate tyrosine residues on proteins. Nitrotyrosine has been shown to be present in proteins from artherosclerotic lesions of human carotid arteries and also in placenta during pre-eclampsia.
Specifications
Nitrotyrosine | |
Monoclonal | |
Unconjugated | |
peroxynitrite-treated KLH | |
RUO | |
Recognizes nitrotyrosine. | |
Purified |
Immunoblot | |
CC22.8C7.3 | |
Mouse | |
50 μg | |
Primary | |
−20°C | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction