Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NK T Cell, Invariant Mouse anti-Human, PE, Clone: 6B11, BD

Catalog No. BDB552825 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552825 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552825 Supplier BD Biosciences Supplier No. 552825
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Monoclonal antibody 6B11 reacts with a unique determinant in the CDR3 region of the invariant (Vα24-JαQ) TCR chain. Thus 6B11 identifies a subset of all Vα24 positive T cells. The binding of the PE-conjugate of 6B11 can be partially blocked by other antibodies specific for Vα24 (BD Biosciences Pharmingen, data not shown). This invariant chain is expressed on invariant NK T cells, which are a small subset of T lymphocytes that also express other NK cell molecules such as CD161. Invariant NK T cells have been reported to play an
immunoregulatory role. The Vα24-JαQ interacts with the CD1d protein. Invariant NK T cells can be specifically activated by CD1d-presented antigen resulting in rapid production of IL-4 and IFN (also known as IFN-γ). Studies of NK T cells describe their role in anti-tumor immunity and autoimmunity. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: 6B11
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen NK T Cell, Invariant
Applications Flow Cytometry
Classification Monoclonal
Clone 6B11
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Peptide from CDR3 loop of the invariant TCR V24 sequence
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.