Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NK-T/NK Cell Antigen (ICAM-1) Rat anti-Mouse, PE, Clone: U5A2-13, BD

Catalog No. BDB550082 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550082 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550082 Supplier BD Biosciences Supplier No. 550082

Rat Monoclonal Antibody

The U5A2-13 mAb recognizes an antigen expressed on most-NKT cells and 55% of NK cells in the liver of various mouse strains, including both NK1.1+ (e.g., C57BL/6) and NK1.1- (e.g., BALB/c, C3H/He, MRL/lpr) strains. Abundant IL-4 and IFN-γ are produced by U5A2-13 Ag+ TCR αβ+ cells upon CD3 cross-linking, but not by U5A2-13 Ag- TCR αβ+ cells in both NK1.1+ and NK1.1- strains. In C57BL/6 mice (NK1.1+ strain), 80%of U5A2-13 Ag+ T cells co-express the NK1.1 molecule. When staining hepatic mononuclear cells with U5A2-13 mAb, the fluorescence intensity detected on on CD3- cells is lower than on CD3+ cells in BALB/c and C57BL/6 mice. Immunoprecipitation studies have shown that the U5A2-13 antigen comprises three proteins of 65, 34, and 31kDa. This antibody clone U5A2-13 was originally established for mouse NKT cells, but recent studies showed that it recognized a novel conformational epitope of ICAM-1.

Host Species: Rat
Clone: U5A2-13
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: BALB/c nu/nu mouse lymphoma line tMK-2U

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen NK-T/NK Cell Antigen (ICAM-1)
Applications Flow Cytometry
Classification Monoclonal
Clone U5A2-13
Concentration 0.2mg/mL
Conjugate PE
Description ICAM-1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen BALB/c nu/nu mouse lymphoma line tMK-2U
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.