Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKB1 Mouse anti-Human, FITC, Clone: DX9, BD

Catalog No. BDB340483 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB340483 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB340483 Supplier BD Biosciences Supplier No. 340483
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-human killer inhibitory receptor (KIR), NKB1, (Anti-Hu KIR [NKB1]) recognizes a 70kDa glycoprotein that is a member of the p58/NK-associated transcript (NKAT) family of class I receptors. KIR (NKB1) is an MHC class I receptor that inhibits NK celln++ated cytotoxicity on target cells that express HLA-B molecules of the Bw4 serotype, but it does not affect recognition of HLA-A or HLA-C antigens. KIR (NKB1)-mediated inhibition is abrogated by binding of Anti-Hu KIR (NKB1). Anti-Hu KIR (NKB1) also regulates T-cell activation initiated through the T-cell antigen receptor (TCR) complex. KIR (NKB1) is expressed on a subset of human NK cells (CD3-/CD56+), NK cell clones, and a subset of human T lymphocytes.

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen NKB1
Applications Flow Cytometry
Classification Monoclonal
Clone DX9
Concentration 1.6μg/mL
Conjugate FITC
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Human NK Cell Clone
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.