Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse NKG2A/CD159a PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody

$139.50 - $467.50

Specifications

Antigen NKG2A/CD159a/KLRC1
Clone 705829
Dilution Flow Cytometry 10 uL/10^6 cells
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FAB6867P025
GREENER_CHOICE
View Documents
R&D Systems
FAB6867P025
25 Tests
Each for $139.50
Only null left
Add to Cart
 
FAB6867P100
GREENER_CHOICE
View Documents
R&D Systems
FAB6867P100
100 Tests
Each for $467.50
Only null left
Add to Cart
 
Description

Description

NKG2A/CD159a/KLRC1 Monoclonal specifically detects NKG2A/CD159a/KLRC1 in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

NKG2A/CD159a/KLRC1
Flow Cytometry 10 uL/10^6 cells
Monoclonal
Purified
RUO
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD159 antigen-like family member A, CD159a, CD159a antigen, C-lectin type II protein, killer cell lectin-like receptor subfamily C, member 1, Klrc1, MGC13374, natural killer cell lectin, natural killer group protein 2, NK cell receptor A, NKG2, NKG2-1/B activating NK receptor, NKG2-A, NKG2-A/B type II integral membrane protein, NKG2-A/B-activating NK receptor, NKG2-A/NKG2-B type II integral membrane protein, NKG2AMGC59791, NKG2-B
KLRC1
IgG2b
Protein A or G purified from hybridoma culture supernatant
Detects mouse NKG2A/CD159a in direct ELISAs.
705829
Flow Cytometry
R-PE
Rat
Mouse
Q9Z202
3821
CHO Chinese hamster ovary cell line transfected with mouse NKG2A/CD159a Accession # Q9Z202
Primary
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.