Learn More
anti-NKG2C/CD159c, Alexa Fluor 700, Clone: 134591, R&D Systems™
Manufacturer:  R&D Systems FAB138N025
 View more versions of this product
Catalog No. FAB138N025
Add to cart
Description
Description
Applications:
Flow Cytometry
Specifications
Specifications
NKG2C/CD159c/KLRC2
134591
Flow Cytometry 5 uL/10^6 cells
Purified
KLRC2
BaF3 mouse pro-B cell line transfected with human NKG2C/CD159c and CD94
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3822
Human
Flow Cytometry
Alexa Fluor 700
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD159 antigen-like family member C, CD159c, CD159c antigen, killer cell lectin-like receptor subfamily C, member 2, KLRC2, MGC138244, NK cell receptor C, NKG2-C, NKG2-C type II integral membrane protein, NKG2CNKG2-C-activating NK receptor
Mouse
IgG1
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects human NKG2C/CD159c heterodimer with CD94 in flow cytometry. It does not cross-react with the human NKG2A/CD94 heterodimer or with the human CD94 homodimer.