Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-NKp46/NCR1, APC, Clone: 29A1.4, R&D Systems™

Catalog No. FAB22252A10
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB22252A10 100 Tests
FAB22252A02 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB22252A10 Supplier R&D Systems Supplier No. FAB22252A100
This item is not returnable. View return policy

Applications:

Flow Cytometry

Specifications

Antigen NKp46/NCR1
Applications Flow Cytometry
Classification Monoclonal
Clone 29A1.4
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q8C567
Gene Alias CD335, CD335 antigen, hNKp46, Ly94, LY94lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46), Lymphocyte antigen 94 homolog, MAR-1, natural cytotoxicity triggering receptor 1, Natural killer cell p46-related protein, NCR1, NK cell-activating receptor, NKp46, NKP46FLJ99094, NK-p46lymphocyte antigen 94 (mouse) homolog (activating NK-receptor; NK-p46)
Gene Symbols NCR1
Host Species Rat
Immunogen Murine myeloma cell line NSO-derived recombinant mouse NKp46-Fc chimera Glu22-Asn255 Accession # Q8C567
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9437
Test Specificity Detects mouse NKp46/NCR1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) NKp30, rhNKp44, rhNKp46, or rhNKp80 is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.