Learn More
NKp46 Mouse anti-Human, PE, Clone: 9E2/NKP46, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 557991
Description
NKp46, a major cytoxicity receptor of NK cells, is a type I membrane glycoprotein (46kDa) is expressed on resting and activated NK cells. Its extracellular region contains two C2-type, Ig-like domains. The transmembrane domain contains a positively charged amino acid (Arg) which could be involved in stabilizing the association with CD3ζ. Its intracellular region does not contain immunoreceptor tyrosine-based activating motifs (ITAM), but it is linked to intracytoplasmic transduction machinery by its association with CD3ζ and FcεRIγ adaptor proteins. NKp46 along with NKp30 and NKp44 are referred to as natural cytoxicity receptors (NCR) and play a very important role in killing virus-infected, tumor cells and MHC-class I-unprotected cells. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: 9E2
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry

Specifications
NKp46 | |
Monoclonal | |
Optimal | |
NCR1; NK-p46; hNKp46; LY94; Natural cytotoxicity triggering receptor 1 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
9E2/NKP46 | |
PE | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human NKp46 Recombinant Protein | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.