Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nkp46 Mouse anti-Human, PE-Cy7, Clone: 9E2/NKp46 (also known as 9-E2), BD

Catalog No. BDB562101 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562101 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562101 Supplier BD Biosciences Supplier No. 562101
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 9E2/Nkp46 monoclonal antibody specifically binds to CD335. CD335 is also known as the Natural killer cell p46-related protein (NKp46) and the Natural cytotoxicity triggering receptor 1 (NCR1). CD335 is a 46 kDa type I membrane glycoprotein that is expressed on resting and activated NK cells. Its extracellular region contains two C2-type, Ig-like domains. The transmembrane domain contains a positively charged amino acid (Arg) which could be involved in stabilizing the association with CD3ζ. Its intracellular region does not contain immunoreceptor tyrosine-based activating motifs (ITAM), but it is linked to intracytoplasmic transduction machinery by its association with CD3ζ and FcεRIγadaptor proteins. CD335 along with NKp30 and NKp44 are referred to as natural cytotoxicity receptors (NCR). These receptors play very important roles in cells that kill virus-infected target cells, tumor cells and MHC-class I-unprotected cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Nkp46
Applications Flow Cytometry
Classification Monoclonal
Clone 9E2/NKp46 (also known as 9-E2)
Conjugate PE-Cyanine7
Description NCR1; NK-p46; hNKp46; LY94; Natural cytotoxicity triggering receptor 1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human NKp46 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.