Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nkx-5.1 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB5744MI Shop All MilliporeSigma Products
missing translation for 'changeView'
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5744MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. AB5744MI Supplier MilliporeSigma missing translation for 'supplierNo' AB5744
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Nkx-5.1 Antibody is validated for use in western blot, immunohistochemistry for the detection of Nkx-5.1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nkx-5.1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Precipitated antibody in a solution of 50% saturated ammonium sulfate and PBS containing no preservatives.
Gene Symbols HMX3; Nkx5-1; HCG1781691; ORFNames=hCG_1781691; NKX5-1
Host Species Rabbit
Immunogen Synthetic peptide, amino acids 10-23 from mouse Nkx5.1.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001105574.1
Target Species Human, Mouse
Content And Storage -70°C for up to 6 months, Avoid Freeze/Thaw Cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.