Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nkx1.1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA535083
Description
The function of this protein remains unknown.
Specifications
| Nkx1.1 | |
| Polyclonal | |
| PBS with 0.09% sodium azide | |
| Q15270 | |
| NKX1-1 | |
| KLH conjugated synthetic peptide between 230-258 amino acids from the central region of human NKX1-1 | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| NKX1-1 | |
| NKX1-1; HPX153; NK1 transcription factor-related protein 1; Homeobox protein 153; Homeobox protein SAX-2; NKX-1.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54729 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction