Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nkx2.1 Mouse anti-Human, Clone: 8G7-G3-1 , MilliporeSigma™

Catalog No. MAB5460MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5460MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5460MI Supplier MilliporeSigma Supplier No. MAB5460
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Nkx2.1 Antibody is validated for use in western blot, immunohistochemistry (paraffin) for the detection of Nkx2.1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nkx2.1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 8G7-G3-1
Conjugate Unconjugated
Formulation Ascites fluid. Liquid. Contains no preservative.
Gene Accession No. NM_001079668.1; NM_003317.3
Gene Symbols NKX2-1, BCH, TEBP, TITF1, TTF1, NKX2.1, NK-2, TTF-1, BHC, NKX2A
Host Species Mouse
Immunogen Recombinant rat Nkx2.1.
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.