Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nkx2.2 Mouse anti-Mouse, Clone: 1C11, MilliporeSigma™

Catalog No. MABD99MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABD99MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABD99MI Supplier MilliporeSigma Supplier No. MABD99

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Anti-Nkx2.2 Antibody, clone 1C11 is a highly specific mouse monoclonal antibody and that targets NKX2.2 and has been tested in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nkx2.2
Applications Western Blot
Classification Monoclonal
Clone 1C11
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_002500
Gene Symbols NKX2-2, NKX2.2, NKX2B
Host Species Mouse
Immunogen Recombinant protein corresponding to human Nkx2.2.
Purification Method Protein G Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.