Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nkx2.2 Mouse, Unlabeled, Clone: 74.5A5, BD

Catalog No. BDB564731 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564731 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564731 Supplier BD Biosciences Supplier No. 564731
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 74.5A5 monoclonal antibody specifically binds to Homeobox protein Nkx-2.2 (Nkx2.2), which is a transcription factor required for the differentiation of ventral neuronal populations in the central nervous system and endocrine cell populations in the pancreas and intestine. Within the pancreas, Nkx2.2 is expressed in many cell subsets and plays important roles in islet and β- cell differentiation and maturation. Mice with a homozygous disruption of the Nkx2-2 gene lack mature β cells and die shortly after birth with severe hyperglycemia. Similarly to the Nkx2.2-ablated mice, homozygous mutations in the human NKX2-2 gene have been described in diabetic patients and associated with the etiology of neonatal diabetes. The NKX2-2 gene is also a target of the Ewing sarcoma specific protein EWS-FLI. Nkx2.2 expression can be monitored in order to determine cell fate changes in cultures for the generation of human pluripotent stem cell-derived insulin-producing cells as well as oligodendrocyte progenitors.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Nkx2.2
Applications Flow Cytometry
Classification Monoclonal
Clone 74.5A5
Concentration 0.5mg/mL
Conjugate Unconjugated
Description NKX2-2; NKX2B; Nkx2-2; Nkx-2.2; NK-2 homolog B; glycoprotein GP330
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Chicken Nkx2.2 Recombinant Protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at 4°C
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.