Learn More
Nkx2.2 Mouse, PE, Clone: 74.5A5, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 564730
Description
The 74.5A5 monoclonal antibody specifically binds to Homeobox protein Nkx-2.2 (Nkx2.2), which is a transcription factor required for the differentiation of ventral neuronal populations in the central nervous system and endocrine cell populations in the pancreas and intestine. Within the pancreas, Nkx2.2 is expressed in many cell subsets and plays important roles in islet and β- cell differentiation and maturation. Mice with a homozygous disruption of the Nkx2-2 gene lack mature β cells and die shortly after birth with severe hyperglycemia. Similarly to the Nkx2.2-ablated mice, homozygous mutations in the human NKX2-2 gene have been described in diabetic patients and associated with the etiology of neonatal diabetes. The NKX2-2 gene is also a target of the Ewing sarcoma specific protein EWS-FLI. Nkx2.2 expression can be monitored in order to determine cell fate changes in cultures for the generation of human pluripotent stem cell-derived insulin-producing cells as well as oligodendrocyte progenitors.

Specifications
Nkx2.2 | |
Monoclonal | |
PE | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Chicken Nkx2.2 Recombinant Protein | |
50 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
74.5A5 | |
NKX2-2; NKX2B; Nkx2-2; Nkx-2.2; NK-2 homolog B; glycoprotein GP330 | |
Mouse | |
Affinity Purified | |
RUO | |
Chicken, Human, Mouse, Rat | |
IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.