Learn More
Nkx6.1 Mouse, Unlabeled, Clone: R11-560, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 563022
Description
The R11-560 monoclonal antibody specifically binds to human and mouse homeobox protein Nkx6.1. The transcription factor Nkx6.1 plays a role in pancreatic islet beta cell development through the regulation of HNF1a and insulin genes. Nkx6.1 is considered to be a highly specific transcription factor marking pancreatic beta islet cells, where it is expressed in both developing and mature cells. In addition, Nkx6.1 functions as a transcriptional repressor induced by the Sonic Hedgehog (Shh) signaling pathway in fate determination of several neuronal cell types. During antibody development, the purified R11-560 monoclonal antibody was found to detect Nkx6.1 by western blot and indirect immunofluorescent staining followed by flow cytometric analysis or imaging analysis.

Specifications
Nkx6.1 | |
Monoclonal | |
0.5mg/mL | |
Nkx6.1, NKX6-1, NKX6A, Nkx6-1, Nkx6.1, Nkx6a | |
Mouse | |
Affinity Purified | |
RUO | |
Human, Mouse | |
IgG1 κ |
Western Blot | |
R11-560 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human Nkx6.1 Recombinant Protein | |
100 μg | |
Primary | |
Store undiluted at 4°C |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.