Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nkx6.3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA535192
Description
This antibody is predicted to react with mouse based on sequence homology.
The NKX family of homeodomain proteins controls numerous developmental processes. Members of the NKX6 subfamily, including NKX6-3, are involved in development of the central nervous system (CNS), gastrointestinal tract, and pancreas.
Specifications
Nkx6.3 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
A6NJ46 | |
NKX6-3 | |
KLH conjugated synthetic peptide between 83-109 amino acids from the central region of human NKX6-3 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
NKX6-3 | |
NKX6-3; Homeobox protein Nkx-6.3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
157848 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction