Learn More
neuroligin 4, Y-linked, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human NLGN4Y protein.
Supplier: Abnova Corporation H00022829D01P
Description
Neuroligins, such as NLGN4Y, are cell adhesion molecules present at the postsynaptic side of the synapse and may be essential for the formation of functional synapses (Jamain et al., 2003 [PubMed 12669065]).[supplied by OMIM
Sequence: MLPIWFTTSLDTLMTYVQDQNEDCLYLNIYVPMEDGTNIKRNADDITSNDHGEDKDIHEQNSKKPVMVYIHGGSYMEGTGNMIDGSILASYGNVIVITINYRLGILGMQEARLCGSSKMFNYFKSPFTNLINFFSpecifications
neuroligin 4, Y-linked | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human NLGN4Y protein. | |
NLGN4Y | |
KIAA0951 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
ENST00000297967 | |
NLGN4Y | |
NLGN4Y (ENSP00000297967, 1 a.a. ∼ 134 a.a) full-length human protein. | |
100 μg | |
Cell Adhesion | |
22829 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.