Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NLRC5 Rat anti-Human, Clone: 3H8, MilliporeSigma™

Catalog No. MABF260MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF260MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF260MI Supplier MilliporeSigma Supplier No. MABF260
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Detect NLRC5 using this rat monoclonal antibody, Anti-NLRC5 Antibody, clone 3H8 validated for use in western blotting, immunoprecipitation and Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NLRC5
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3H8
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols NLRC5, NOD27, NOD4
Host Species Rat
Immunogen BSA-conjugated linear peptide corresponding to the C-terminus of human NLRC5.
Purification Method Affinity
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.