Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NLRP11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
Antigen | NLRP11 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Immunohistochemistry-Paraffin 5 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224688SS
![]() |
Novus Biologicals
NBP224688SS |
0.025 mL |
Each for $181.90
|
|
|||||
NBP224688
![]() |
Novus Biologicals
NBP224688 |
0.1 mL |
Each for $402.00
|
|
|||||
Description
NLRP11 Polyclonal specifically detects NLRP11 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
NLRP11 | |
Immunohistochemistry-Paraffin 5 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
CLR19.6, NACHT, leucine rich repeat and PYD containing 11, NALP11NACHT, LRR and PYD domains-containing protein 11, NLR family, pyrin domain containing 11, NOD17PYPAF7, Nucleotide-binding oligomerization domain protein 17, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 11, PAAD- and NACHT-containing protein 10, PAAD- and NACHT-containing protein 10B, PAAD-and NACHT domain-containing protein 10, PAN10FLJ26273, PYPAF6NACHT, LRR and PYD containing protein 11, PYRIN-containing APAF1-like protein 6 | |
NLRP11 | |
IgG | |
Protein A purified | |
This antibody will detect the approximately 300 kD full-length Notch1form and the approximately 100 kD cleaved Notch1 form of the protein. |
0.5 mg/mL | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
P59045 | |
204801 | |
A portion of amino acids 850-900 of human NALP11 was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title