Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NMDA R, NR1 Subunit Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB300115
Description
NMDA R, NR1 Subunit Polyclonal specifically detects NMDA R, NR1 Subunit in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
NMDA receptor C1 | |
Polyclonal | |
Unconjugated | |
No Preservative | |
Rabbit | |
120 kDa | |
0.025 mg | |
Neuronal Cell Markers, Neuroscience, Neurotransmission | |
Specific for the ∼120k NR1 subunit of the NMDA receptor containing the C1 splice variant insert. Does not recognize the NR1 subunits of the NMDA receptor that do not contain the C1 insert. | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
LYOPH | |
Western Blot 1:1000, Immunohistochemistry 1:1000-1:2000, Immunohistochemistry-Frozen 1:1000-1:2000 | |
GluN1, MRD8, NDHMSD, NMDA1, NMDAR1, NR1 | |
Synthetic peptide corresponding to amino acid residues specific to the NR1 subunit, C1 splice variant insert conjugated to KLH. Accession # P35439 | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Reconstitute with 50 ul PBS to desired concetration. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction