Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-NMDAR 2B (984-1104), Polyclonal

Catalog No. 45458210UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
45-458-210UG 10 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-458-210UG Supplier MilliporeSigma™ Supplier No. 45458210UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects NMDAR 2B (984-1104) Clone: in Fish, Human, Mouse, Rat samples, and it is validated for Immunohistochemistry (Frozen), Immunoprecipitation, Immuno Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen NMDAR 2B (984-1104)
Applications Immunoblot, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation Lyophilized from 5mM ammonium bicarbonate.
Host Species Rabbit
Immunogen a recombinant fusion protein consisting of amino acids 984-1104 of rat brain NMDAR 2B-subunit
Quantity 10 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽180kDa NMDA receptor NR2B subunit.
Target Species Fish, Human, Mouse, Rat
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.