Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMDAR1 Rabbit anti-Rat, Clone: 1.17.2.6, MilliporeSigma™

Catalog No. AB9864MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9864MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9864MI Supplier MilliporeSigma Supplier No. AB9864
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

This Anti-NMDAR1 Antibody is validated for use in immunohistochemistry, western blotting for the detection of NMDAR1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMDAR1
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 1.17.2.6
Conjugate Unconjugated
Formulation Rabbit Monoclonal supernatant liquid containing 0.05% sodium azide.
Gene Accession No. NM_000832.5
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminus of rat NMDA receptor subunit (Catalog number AG344). {amino acids 909-938 rat NMDAR1{LQNQKDTVLPRRAIERE EGQLQLCSRHRES}
Purification Method Unpurified
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) GRIN1, NR1, Glutamate, NMDAR, NMDA1, NMDAR1
Target Species Rat
Content And Storage −20°C for 12 months
Form Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.