Learn More
NMDAR1 Splice Variant C1 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | NMDAR1 Splice Variant C1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Lyophilized from ammonium bicarbonate (5 mM) so some residual salt may also be present. Reconstitute with 50μL of PBS. Contains no preservatives. |
| Gene Symbols | GRIN1; Glutamate; NMDAR1; NMDA1; NR1 |
| Host Species | Rabbit |
| Immunogen | Peptide corresponding to the C-terminus of the rat NMDAR1 C1 splice variant. |
| Purification Method | Affinity Purified |
| Quantity | 25 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.